MilliporeSigma
  • Proteomic profiling of mouse epididymosomes reveals their contributions to post-testicular sperm maturation.
Proteomic profiling of mouse epididymosomes reveals their contributions to post-testicular sperm maturation.
Molecular & cellular proteomics : MCP (2018-09-15)
Brett Nixon, Geoffry N De Iuliis, Hanah M Hart, Wei Zhou, Andrea Mathe, Ilana Bernstein, Amanda L Anderson, Simone J Stanger, David A Skerrett-Byrne, M Fairuz B Jamaluddin, Juhura G Almazi, Elizabeth G Bromfield, Martin R Larsen, Matthew D Dun
ABSTRACT
The functional maturation of spermatozoa that is necessary to achieve fertilization occurs as they transit through the epididymis, a highly specialized region of the male reproductive tract. A defining feature of this maturation process is that it occurs in the complete absence of nuclear gene transcription or de novo protein translation in the spermatozoa. Rather, it is driven by sequential interactions between spermatozoa and the complex external milieu in which they are bathed within lumen of the epididymal tubule. A feature of this dynamic microenvironment are epididymosomes, small membrane encapsulated vesicles that are secreted from the epididymal soma. Herein, we report comparative proteomic profiling of epididymosomes isolated from different segments of the mouse epididymis using multiplexed tandem mass tag (TMT) based quantification coupled with high resolution LC-MS/MS. A total of 1640 epididymosome proteins were identified and quantified via this proteomic method. Notably, this analysis revealed pronounced segment-to-segment variation in the encapsulated epididymosome proteome. Thus, 146 proteins were identified as being differentially accumulated between caput and corpus epididymosomes, and a further 344 were differentially accumulated between corpus and cauda epididymosomes (i.e. fold change of  -1.5 or  1.5; P < 0.05). Application of gene ontology annotation revealed a substantial portion of the epididymosome proteins mapped to the cellular component of extracellular exosome and to the biological processes of transport, oxidation-reduction, and metabolism. Additional annotation of the subset of epididymosome proteins that have not previously been identified in exosomes revealed enrichment of categories associated with the acquisition of sperm function (e.g. fertilization and binding to the zona pellucida). In tandem with our demonstration that epididymosomes are able to convey protein cargo to the head of maturing spermatozoa, these data emphasize the fundamental importance of epididymosomes as key elements of the epididymal microenvironment responsible for coordinating post-testicular sperm maturation.
Sigma-Aldrich
Anti-PDIA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution