Skip to Content
MilliporeSigma
Triisopropylsilyl trifluoromethanesulfonate 97%

Triisopropylsilyl trifluoromethanesulfonate

Triisopropylsilyl trifluoromethanesulfonate 97%
Synonym(s):
TIPS triflate, Trifluoromethanesulfonic acid triisopropylsilyl ester, Triisopropylsilyl triflate
Linear Formula:
CF3SO3Si[CH(CH3)2]3
CAS Number:
Molecular Weight:
306.42

Products

Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing