Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248460

Sigma-Aldrich

Triisopropylsilyl trifluoromethanesulfonate

97%

Synonym(s):

TIPS triflate, Trifluoromethanesulfonic acid triisopropylsilyl ester, Triisopropylsilyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si[CH(CH3)2]3
CAS Number:
Molecular Weight:
306.42
Beilstein:
3591541
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.415 (lit.)

bp

45-46 °C/0.03 mmHg

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C

InChI

1S/C10H21F3O3SSi/c1-7(2)18(8(3)4,9(5)6)16-17(14,15)10(11,12)13/h7-9H,1-6H3

InChI key

LHJCZOXMCGQVDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare silyloxy acetylenes from lithium acetylides; the silyloxy acetylenes were then employed in protic acid-promoted cyclizations of arenes, alkynes and olefins.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryunosuke Kuriki et al.
Dalton transactions (Cambridge, England : 2003), 49(35), 12234-12241 (2020-08-28)
New types of diaryloxystannylenes and -plumbylenes have been synthesized and structurally characterized. Lappert's diaminostannylene and -plumbylene E[N(SiMe3)2]2 (E = Sn, Pb) react with 1,3-diethers of tetrathiacalix[4]arene bearing benzyl or SiiPr3 groups to give the corresponding diaryloxystannylenes and -plumbylenes embedded in
Greene, T.W. Wuts, P.G.M.
Protective Groups in Organic Synthesis, 74-74 (1991)
Synlett, 435-435 (1993)
Tetrahedron, 62, 11371-11371 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service