Skip to Content
MilliporeSigma
Triethylsilane 99%

Triethylsilane

Triethylsilane 99%
Synonym(s):
NSC 93579, Triethylhydrosilane, Triethylsilicon hydride, Triethylsilane
Linear Formula:
(C2H5)3SiH
CAS Number:
Molecular Weight:
116.28

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing