Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18806

Sigma-Aldrich

Triethylsilane

for synthesis

Synonym(s):

Triethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiH
CAS Number:
Molecular Weight:
116.28
MDL number:
UNSPSC Code:
12352103
EC Index Number:
210-535-3
NACRES:
NA.22

vapor pressure

>1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

reaction suitability

reagent type: reductant

bp

107-109 °C/1013 hPa

transition temp

flash point -6 °C

density

0.73 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](CC)(CC)CC

InChI

1S/C6H15Si/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

QXTIBZLKQPJVII-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.730 - 0.732
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F

Flash Point(C)

-6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service