Tricine
Synonym(s):
N-[Tris(hydroxymethyl)methyl]glycine, Tricine, Tricine, ULTROL® Grade
Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing