Skip to Content
MilliporeSigma
Sodium thiosulfate pentahydrate ACS reagent, ≥99.5%

Sodium thiosulfate pentahydrate

Sodium thiosulfate pentahydrate ACS reagent, ≥99.5%
Synonym(s):
Sodium thiosulfate pentahydrate
Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade