Skip to Content
MilliporeSigma
Sodium 1-pentanesulfonate monohydrate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)

Sodium 1-pentanesulfonate monohydrate

Sodium 1-pentanesulfonate monohydrate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)
Synonym(s):
1-Pentanesulfonic acid sodium salt
Linear Formula:
CH3(CH2)4SO3Na · H2O
CAS Number:
Molecular Weight:
192.21
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique