Skip to Content
MilliporeSigma
Poly(L-lactide) viscosity ~4.0 dL/g, 0.1 % (w/v) in chloroform(25 °C)

Poly(L-lactide)

Poly(L-lactide) viscosity ~4.0 dL/g, 0.1 % (w/v) in chloroform(25 °C)
Synonym(s):
L-Lactide polymer, PLA, Poly(L-Lactic acid), PLA, Poly(L-Lactic acid)
Linear Formula:
CH3O(C6H8O4)nH

Products

Brand
Physical Form
Quality Segment
Optical Isomer
Polymer Composition
Polymer Type