Skip to Content
MilliporeSigma
Methyl 3-chloro-3-oxopropionate 97%

Methyl 3-chloro-3-oxopropionate

Methyl 3-chloro-3-oxopropionate 97%
Synonym(s):
Methyl malonyl chloride, Methyl chloroformylacetate
Linear Formula:
CH3OCOCH2COCl
CAS Number:
Molecular Weight:
136.53
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing