Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164011

Sigma-Aldrich

Methyl 3-chloro-3-oxopropionate

97%

Synonym(s):

Methyl malonyl chloride, Methyl chloroformylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2COCl
CAS Number:
Molecular Weight:
136.53
Beilstein:
1754078
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.432 (lit.)

bp

57-59 °C/12 mmHg (lit.)

density

1.273 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)CC(Cl)=O

InChI

1S/C4H5ClO3/c1-8-4(7)2-3(5)6/h2H2,1H3

InChI key

UTBCRHAMJFMIIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 3-chloro-3-oxopropionate reacts with 4-cyano-1,1′-biphenyl derivatives bearing ω-hydroxyalkyl substituents to yield liquid-crystalline linear malonates and cyanoacetates.

Application

Methyl 3-chloro-3-oxopropionate was used as a reagent in the synthesis of resin-bound 4-tosyl quinolinone, 1-anthracen-9-ylmethyl 3-methyl 2,2-dimethylmalonate and 1-(4-(3,5-dihydropyren-1-yl)butyl)-3-methyl 2,2-dimethylmalonate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general solid phase synthesis of 4-substituted quinolinones via Pd-catalyzed cross coupling.
Xu C, et al.
Tetrahedron Letters, 47(28), 4885-4888 (3006)
Solvent-free microwave-assisted Bingel reaction in carbon nanohorns.
Economopoulos SP, et al.
Journal of Materials Chemistry, 19(39), 7326-7331 (2009)
Katharina C Kress et al.
Beilstein journal of organic chemistry, 8, 371-378 (2012-04-18)
4-Cyano-1,1'-biphenyl derivatives bearing ω-hydroxyalkyl substituents were reacted with methyl 3-chloro-3-oxopropionate or cyanoacetic acid, giving liquid-crystalline linear malonates and cyanoacetates. These compounds formed monotropic nematic phases at 62 °C down to ambient temperature upon cooling from the isotropic liquid. The mesomorphic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service