Skip to Content
MilliporeSigma
Ethyl octanoate ≥98%, FCC, FG

Ethyl octanoate

Ethyl octanoate ≥98%, FCC, FG
Synonym(s):
Ethyl caprylate, Ethyl octanoate
Linear Formula:
CH3(CH2)6COOC2H5
CAS Number:
Molecular Weight:
172.26

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4