Skip to Content
MilliporeSigma
All Photos(1)

Documents

W244902

Sigma-Aldrich

Ethyl octanoate

≥98%, FCC, FG

Synonym(s):

Ethyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOC2H5
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2449
Beilstein:
1754470
EC Number:
Council of Europe no.:
392
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.111
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

0.02 mmHg ( 25 °C)

Assay

≥98%

refractive index

n20/D 1.417 (lit.)

bp

206-208 °C (lit.)

mp

−48-−47 °C (lit.)

density

0.867 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

waxy; fruity; pineapple; sweet

SMILES string

CCCCCCCC(=O)OCC

InChI

1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3

InChI key

YYZUSRORWSJGET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl octanoate is a volatile compound reported to be found in Chinese rice wine and ripe ‘Kensington Pride′ mango fruit.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Iwamoto et al.
Asia Pacific journal of clinical nutrition, 23(2), 256-262 (2014-06-06)
Serum undercarboxylated osteocalcin (ucOC) is an index of vitamin K nutritional status in treatment-naive postmenopausal osteoporotic women. The purpose of the present study was to reveal the association between vitamin K nutritional status and serum ucOC concentrations in postmenopausal osteoporotic
Aroma volatiles production during fruit ripening of `Kensington Pride?mango.
Postharvest biology and technology, 27(3), 323-336 (2003)
The influence of yeast strains on the volatile flavour compounds of Chinese rice wine
Chen S and Xu Y
Journal of the Institute of Brewing, 116(2), 190-196 (2010)
Silawan Somboonchan et al.
Food chemistry, 195, 79-86 (2015-11-18)
The effect of modulating the gelatinisation extent by hydration (50/50 and 80/20 water to starch ratio) and temperature (65 or 85 °C) on various properties of wheat starch in presence of flavours has been studied. The hydrothermal treatments resulted in
Marwa Ahmed Sallam et al.
International journal of nanomedicine, 10, 4459-4478 (2015-07-24)
In this study, an optimized nanodispersible oral dosage form (containing a lactate ester) was developed for cilostazol (CZL). CZL is a phosphodiesterase inhibitor used for intermittent claudication. We aimed to improve the dissolution rate and absorption of CZL giving it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service