Skip to Content
MilliporeSigma
Esmolol hydrochloride United States Pharmacopeia (USP) Reference Standard

Esmolol hydrochloride

Esmolol hydrochloride United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzenepropanoic acid methyl ester hydrochloride hydrochloride
Empirical Formula (Hill Notation):
C16H26NClO4
CAS Number:
Molecular Weight:
331.83

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2