Skip to Content
MilliporeSigma
D-Glucose-1,2,3,4,5,6,6-d7 endotoxin tested, 97 atom % D

D-Glucose-1,2,3,4,5,6,6-d7

D-Glucose-1,2,3,4,5,6,6-d7 endotoxin tested, 97 atom % D
Synonym(s):
D-Glucose-C-d7, Dextrose-C-d7
Empirical Formula (Hill Notation):
C6H5D7O6
CAS Number:
Molecular Weight:
187.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing