Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706663

Sigma-Aldrich

D-Glucose-1,2,3,4,5,6,6-d7

endotoxin tested, 97 atom % D

Synonym(s):

Deuterated Glucose, Labeled Glucose, D-Glucose-C-d7, Dextrose-C-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5D7O6
CAS Number:
Molecular Weight:
187.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

≥99% (HPLC)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (dec) (lit.)

suitability

endotoxin tested

mass shift

M+7

SMILES string

[2H]C([2H])(O)[C@@]1([2H])OC([2H])(O)[C@]([2H])(O)[C@@]([2H])(O)[C@]1([2H])O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4-,5+,6?/m0/s1/i1D2,2D,3D,4D,5D,6D

InChI key

WQZGKKKJIJFFOK-KMTOWNPSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the enduser.

Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Zhang et al.
Plant biotechnology journal, 19(12), 2517-2531 (2021-08-04)
Leaf angle is one of the key factors that determines rice plant architecture. However, the improvement of leaf angle erectness is often accompanied by unfavourable changes in other traits, especially grain size reduction. In this study, we identified the pow1
Chunmei Yang et al.
International journal of biological sciences, 16(6), 1059-1070 (2020-03-07)
Cullin 4A and 4B (CUL4A and 4B) function as oncogenes in colorectal cancer (CRC) cells. Both of them conservatively associate with DNA damage-binding protein 1 (DDB1) and DDB1-CUL4-associated factor 4 (DCAF4) to form Cullin-RING E3 ligases known as CRL4DCAF4, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service