Skip to Content
MilliporeSigma
Calcium nitrate tetrahydrate JIS special grade, ≥98.5%

Calcium nitrate tetrahydrate

Calcium nitrate tetrahydrate JIS special grade, ≥98.5%
Synonym(s):
Calcium nitrate tetrahydrate
Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Technique