Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02120

Supelco

Calcium nitrate tetrahydrate

EMPLURA®

Synonym(s):

Calcium nitrate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4 H2O
CAS Number:
Molecular Weight:
236.15
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-332-1
NACRES:
NA.21
form:
solid
solubility:
2710 g/L

Quality Level

product line

EMPLURA®

Assay

≥98.0% (complexometric)

form

solid

potency

3900 mg/kg LD50, oral (Rat)

impurities

≤0.3% Substances not precipitated by ammonium oxalate (as sulfate)

pH

5.0-7.0 (25 °C, 50 g/L in H2O)

mp

45 °C

solubility

2710 g/L

density

1.82 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.001%
NH4+: ≤0.005%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[Ca+2].[N+](=O)([O-])O.[N+](=O)([O-])O.O.O.O.O

InChI

1S/Ca.2HNO3.4H2O/c;2*2-1(3)4;;;;/h;2*(H,2,3,4);4*1H2/q+2;;;;;;

InChI key

DKCJOKYHYIAZQH-UHFFFAOYSA-N

Application


  • Influence of impurity on the properties of chemically synthesized calcium hydroxide: This study utilized Calcium nitrate tetrahydrate (Ca(NO3)2.4H2O) for the chemical precipitation synthesis of calcium hydroxide at room temperature (P Kumar et al., 2021).

  • Study of the effect of calcium nitrate, calcium formate, triethanolamine, and triisopropanolamine on compressive strength of Portland-pozzolana cement: Explored the impact of Calcium nitrate tetrahydrate on the performance of Portland-pozzolana cement (A Derakhshani et al., 2023).

  • Calcium nitrate tetra-hydrate-doped polyvinylidene fluoride thin films as potential devices for pressure sensors: Discusses the preparation and potential use of these doped films in sensor applications (KS Hwang et al., 2020).

  • Effect of reaction rate on the properties of chemically synthesized calcium hydroxide nanoparticles: Investigates how varying the reaction rate affects the properties of nanoparticles synthesized using Calcium nitrate tetrahydrate (P Kumar et al., 2020).

Analysis Note

Assay (complexometric): ≥ 98.0 %
pH-value (5 %; water): 4.0 - 6.0
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.02 %
Heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.005 %
Substances not precipitated by ammonium oxalate (as sulfate): ≤ 0.3 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service