Skip to Content
MilliporeSigma
Benzyltriethylammonium chloride 99%

Benzyltriethylammonium chloride

Benzyltriethylammonium chloride 99%
Synonym(s):
Benzyltriethylammonium chloride
Linear Formula:
C6H5CH2(C2H5)3NCl
CAS Number:
Molecular Weight:
227.77

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing