
Adenosine A1 Receptor Antagonist I

Synonym(s):
8-Cyclopentyl-1,3-dipropylxanthine, 1,3-Dipropyl-8-cyclopentylxanthine, DPCPX, PD 116,948
Empirical Formula (Hill Notation):
C16H24N4O2
CAS Number:
Molecular Weight:
304.39
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing