Skip to Content
MilliporeSigma

119147

Adenosine A1 Receptor Antagonist I

DPCPX

Synonym(s):

8-Cyclopentyl-1,3-dipropylxanthine, 1,3-Dipropyl-8-cyclopentylxanthine, DPCPX, PD 116,948

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H24N4O2
CAS Number:
Molecular Weight:
304.39
UNSPSC Code:
12352200
NACRES:
NA.51
MDL number:
Form:
white solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

white solid

solubility

soluble (DMSO (5 mM))

storage temp.

2-8°C

SMILES string

CCCN1C(=O)N(CCC)c2nc([nH]c2C1=O)C3CCCC3

InChI

1S/C16H24N4O2/c1-3-9-19-14-12(15(21)20(10-4-2)16(19)22)17-13(18-14)11-7-5-6-8-11/h11H,3-10H2,1-2H3,(H,17,18)

InChI key

FFBDFADSZUINTG-UHFFFAOYSA-N

General description

A blood-brain barrier permeable xanthine derivative that acts as a highly selective antagonist of Adenosine A1 receptor (A1R; Ki = 3.9 nM, 130 nM, 1.0 µM, 4.0 µM for human A1, A2A, A2B, and A3 respectively). Exhibits about 1,000-fold higher affinity and a 30-fold greater selectivity for A1R when compared to theophylline. Shown to competitively antagonize the inhibition of adenylate cyclase activity via A1 adenosine receptors in rat fat cells and the stimulation of adenylate cyclase via A2 adenosine receptors in human platelets. Exhibits hypertensive properties.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library