Skip to Content
MilliporeSigma
ADA BioUltra, ≥99.0% (T)

ADA

ADA BioUltra, ≥99.0% (T)
Synonym(s):
N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid
Linear Formula:
H2NCOCH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
190.15
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing