Skip to Content
AS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Testing
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon disulfide
Synonym(s):
Carbon Disulfide, Carbon disulfide
Empirical Formula (Hill Notation):
CS
2
CAS Number:
75-15-0
Molecular Weight:
76.14
EC Number:
200-843-6
Products
Product Category
solvents (10)
Brand
Sigma-Aldrich (8)
Supelco (8)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (3)
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
180173
Carbon disulfide,
ACS reagent, ≥99.9%
Expand
View Pricing
270660
Carbon disulfide,
suitable for HPLC, ≥99.9%
Expand
View Pricing
342270
Carbon disulfide,
ReagentPlus
®
, low benzene, ≥99.9%
Expand
View Pricing
676918
Carbon disulfide,
ACS reagent, ≥99.9%
Expand
View Pricing
CX0397
Carbon disulfide,
low benzene OmniSolv
®
Expand
View Pricing
1.02214
Carbon disulfide,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
31627
Carbon disulfide,
suitable for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC)
Expand
View Pricing
1.02211
Carbon disulfide,
EMPLURA
®
Expand
View Pricing
335266
Carbon disulfide,
anhydrous, ≥99%
Expand
View Pricing
424641
Carbon disulfide,
ReagentPlus
®
, purified by redistillation, ≥99.9%
Expand
View Pricing
1.02210
Carbon disulfide,
for spectroscopy Uvasol
®
Expand
View Pricing
1.94513
Carbon disulfide,
EMPLURA
®
Expand
View Pricing
CX0396
Carbon disulfide,
Meets Reagent Specifications for testing USP/NF monographs OmniSolv
®
Expand
View Pricing
CX0395
Carbon disulfide,
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
PHR3503
Carbon disulfide,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
05-1560
Carbon disulfide,
CP
Expand
View Pricing