Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02211

Supelco

Carbon disulfide

EMPLURA®

Synonym(s):

Carbon disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12191602
EC Index Number:
200-843-6
NACRES:
NA.21
bp:
46.5 °C/1013 hPa
vapor pressure:
398 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

398 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

95 °C

potency

3188 mg/kg LD50, oral (Rat)

expl. lim.

1-60 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.0005% Sulphate (SO4)
≤0.003% Sulfite (as SO2)
≤0.02% Water (Karl Fischer)

non-volatile residue (NVR)

≤0.005%

bp

46.5 °C/1013 hPa

mp

-111.6 °C

transition temp

flash point -30 °C

density

1.26 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Multi-omics analysis of nitrifying sludge under carbon disulfide stress: Nitrification performance and molecular mechanisms.: This study presents a multi-omics analysis of nitrifying sludge under carbon disulfide stress, detailing impacts on nitrification performance and molecular mechanisms, providing insights into environmental and industrial applications of carbon disulfide in waste management and biological treatment processes (Liu et al., 2024).

  • Design and synthesis of novel 1,3,4-thiadiazole thioglycosides as promising antimicrobial potent structures.: Highlights the synthesis of 1,3,4-thiadiazole thioglycosides using carbon disulfide, exploring their antimicrobial potential, which may contribute to the development of new treatments for bacterial infections, particularly in resistance scenarios (Abu-Zaied et al., 2024).

  • New Insights on Solvent-Induced Changes in Refractivity and Specific Rotation of Poly(propylene oxide) Systems Extracted from Channeled Spectra.: Discusses findings on how solvents influence the optical properties of polymeric systems involving carbon disulfide, relevant to materials science and chemical engineering research (Zara et al., 2024).

Analysis Note

Assay (GC): ≥ 99.5 %
Identity (IR): passes test
Water (K. F.): ≤ 0.02 %
Sulphate (SO₄): ≤ 0.0005 %
Non-volatile matter: ≤ 0.005 %
Hydrogensulfid and foreign organic sulfur compounds: passes test
Sulfite (as SO₂): ≤ 0.003 %
Conforms to ÖAB

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service