Skip to Content
MilliporeSigma
4′-Chloroacetanilide 97%

4′-Chloroacetanilide

4′-Chloroacetanilide 97%
Synonym(s):
N-(4-Chlorophenyl)acetamide, Acetic acid 4-chloroanilide, NSC 40563, NSC 444, 4-Chloroacetanilide
Linear Formula:
CH3CONHC6H4Cl
CAS Number:
Molecular Weight:
169.61

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing