Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41480

Sigma-Aldrich

4-Chloroacetanilide

for synthesis

Synonym(s):

4-Chloroacetanilide, N-(4-chlorophenyl)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8ClNO
CAS Number:
Molecular Weight:
169.61
MDL number:
UNSPSC Code:
12352111
EC Index Number:
208-707-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

333 °C/1013 hPa

mp

179-180 °C

solubility

0.95 g/L (experimental)

density

1.385 g/cm3 at 22 °C

storage temp.

2-30°C

SMILES string

Clc1ccc(cc1)NC(=O)C

InChI

1S/C8H8ClNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)

InChI key

GGUOCFNAWIODMF-UHFFFAOYSA-N

Application

<ul>
<li><strong>High-performance liquid chromatography (HPLC) method validation:</strong> 4 Chloroacetanilide is used as an internal standard for isosorbide dinitrate in the assay of sustained-release tablets or capsules containing nitroglycerin, isosorbide dinitrate, or pentaerythritol tetranitrate by high-performance liquid chromatography (Gelber and Papas, 1983).</li>
</ul>

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 177 °C
Melting range (upper value): ≤ 180 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service