Skip to Content
MilliporeSigma
2-Naphthyl phosphate sodium salt

2-Naphthyl phosphate sodium salt

2-Naphthyl phosphate sodium salt
Synonym(s):
β-Naphthyl phosphate monosodium salt, 2-Naphthyl phosphate monosodium salt, mono-Sodium 2-naphthyl phosphate sodium salt
Empirical Formula (Hill Notation):
C10H8NaO4P
CAS Number:
Molecular Weight:
246.13
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing