Skip to Content
MilliporeSigma
(1R)-(−)-Nopol 98%

(1R)-(−)-Nopol

(1R)-(−)-Nopol 98%
Synonym(s):
(−)-Nopol, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol, 6,6-Dimethyl-2-norpinene-2-ethanol, 6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol
Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing