Skip to Content
MilliporeSigma
All Photos(1)

Documents

340367

Sigma-Aldrich

(1R)-(−)-Nopol

98%

Synonym(s):

(−)-Nopol, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol, 6,6-Dimethyl-2-norpinene-2-ethanol, 6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
Beilstein:
3196379
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

viscous liquid

optical activity

[α]24/D −37°, neat

refractive index

n20/D 1.493 (lit.)

bp

230-240 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CC1(C)[C@H]2CC=C(CCO)[C@@H]1C2

InChI

1S/C11H18O/c1-11(2)9-4-3-8(5-6-12)10(11)7-9/h3,9-10,12H,4-7H2,1-2H3/t9-,10-/m0/s1

InChI key

ROKSAUSPJGWCSM-UWVGGRQHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1R)-(-)-Nopol is a chiral terpenol derivative.

Application

(1R)-(-)-Nopol may be used in the preparation of optically active O,O-diterpenyl dithiophosphoric and O-terpenyl aryldithiophosphonic acids by reacting with tetraphosphorus decasulfide and 2,4-diaryl 1,3,2,4-dithiadiphosphetane-2,4-disulfides, respectively. It may also react with silver hexafluoroantimonate to form a complex, which can be an effective initiator for the nonthermal curing of epoxy resins by electron beam induced cationic polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Silver olefin complexes: highly efficient initiators for the electron beam curing of epoxy resins.
Barriau E, et al.
Macromolecules, 41(11), 3779-3781 (2008)
Terpene Analogues of Dithiophospate Pesticides.
Cherkasov RA, et al.
Phosph. Sulfur Relat. Elem., 186(4), 1003-1004 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service