Skip to Content
MilliporeSigma
1-Decanol analytical standard

1-Decanol

1-Decanol analytical standard
Synonym(s):
n-Decyl alcohol, Alcohol C10, 1-Decanol
Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing