
1,5-Diamino-2-methylpentane

Synonym(s):
2-Methyl-1,5-pentanediamine, 2-Methylpentamethylene diamine
Linear Formula:
NH2CH2CH2CH2CH(CH3)CH2NH2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing