Skip to Content
MilliporeSigma

1699016

USP

Tropicamide Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(Ethylaminomethyl)pyridine, N-(Pyridin-4-ylmethyl)ethanamine, N-Ethyl-4-pyridinemethanamine, NSC 85903

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

tropicamide

manufacturer/tradename

USP

refractive index

n20/D 1.516 (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCNCc1ccncc1

InChI

1S/C8H12N2/c1-2-9-7-8-3-5-10-6-4-8/h3-6,9H,2,7H2,1H3

InChI key

ZBAMQLFFVBPAOX-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tropicamide Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Tropicamide

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​


Still not finding the right product?

Explore all of our products under Tropicamide Related Compound A


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library