Skip to Content
MilliporeSigma

Skip To

1683606

USP

Triethyl citrate

United States Pharmacopeia (USP) Reference Standard

Sustainability features

Synonym(s):

Ethyl citrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1801199
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

API family

triethyl citrate

form

liquid

manufacturer/tradename

USP

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product, Sustainability features

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)
pharmaceutical

format

neat

greener alternative category

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Triethyl citrate is used in microbiology primarily as a plasticizer in the formulation of biologics, where it enhances the flexibility of polymeric materials. In biologic product development, it is also used as a stabilizing agent in vaccines and other injectable therapeutics. Its role in drug formulations is crucial for maintaining the integrity and performance of biologic products over time.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with "Less Hazardous Chemical Syntheses". Click here for more information.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Ph

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1153W308302D1300000
description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

natural, ≥99%, FG

description

European Pharmacopoeia (EP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical secondary standard, certified reference material

grade

Halal, natural, Fragrance grade, FG, Kosher

grade

pharmaceutical primary standard

form

liquid

form

-

form

-

form

-

format

neat

format

neat

format

-

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

flavors and fragrances

application(s)

pharmaceutical (small molecule)

vapor density

9.7 (vs air)

vapor density

9.7 (vs air)

vapor density

9.7 (vs air)

vapor density

-


Still not finding the right product?

Explore all of our products under Triethyl citrate


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

311.0 °F - closed cup

Flash Point (°C)

155 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. (1) What will be purity of the Triethyl Citrate? (2) How much Diethyl Phthalate (impurity) will be at ppm level in this Triethyl Citrate?

    1 answer
    1. This product is a product of the United States Pharmacopeia. The material is a neat or pure liquid listed as a 996ug/mg purity by the supplier. The impurities are not identified.

      Please see the link below to review the supplier product page.
      https://store.usp.org/product/1683606?_gl=1*lyrcq3*_gcl_au*NDA2MDgyNjYuMTY5Njk2MTEyMA..*_ga*MTg3MzY4OTg2Ny4xNjg2NTg2MTE5*_ga_DTGQ04CR27*MTcwMDUxNTA5OS42LjAuMTcwMDUxNTA5OS4wLjAuMA..

      Helpful?

Reviews

No rating value

Active Filters