Skip to Content
MilliporeSigma

Skip To

1613860

USP

Sodium cyclamate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium N-cyclohexylsulfamate, N-Cyclohexylsulfamic acid sodium salt, Cyclamic acid sodium salt, Cyclohexanesulfamic acid sodium salt, Sodium cyclamate, Sodium cyclohexanesulfamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H11NHSO3Na
CAS Number:
Molecular Weight:
201.22
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4166868
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sodium cyclamate

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[O-]S(=O)(=O)NC1CCCCC1

InChI

1S/C6H13NO3S.Na/c8-11(9,10)7-6-4-2-1-3-5-6;/h6-7H,1-5H2,(H,8,9,10);/q;+1/p-1

InChI key

UDIPTWFVPPPURJ-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium cyclamate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Phenylsulfamate sweet tastant detectable by humans. ′First generation′ sweetener studied in animal models for its carcinogenic potential.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S076000047827PHR3740
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

pharmaceutical

format

neat

format

neat

format

neat

format

-

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

-

API family

sodium cyclamate

API family

sodium cyclamate

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Sodium cyclamate


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters