Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1563003

USP

Probenecid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

p-(Dipropylsulfamoyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19NO4S
CAS Number:
Molecular Weight:
285.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

probenecid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(O)=O

InChI

1S/C13H19NO4S/c1-3-9-14(10-4-2)19(17,18)12-7-5-11(6-8-12)13(15)16/h5-8H,3-4,9-10H2,1-2H3,(H,15,16)

InChI key

DBABZHXKTCFAPX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Probenecid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Probenecid and Colchicine Tablets
  • Probenecid Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical pharmacokinetics of probenecid.
Cunningham, R. F., Z. H. Israili, and P. G. Dayton.
Clinical Pharmacokinetics, 6.2, 135-151 (1981)
Probenecid and zidovudine metabolism.
Kornhauser, DavidM, et al.
Lancet, 334.8661, 473-475 (1989)
R F Cunningham et al.
Clinical pharmacokinetics, 6(2), 135-151 (1981-03-01)
A review of the clinical applications and of the disposition of probenecid in man, including drug interactions, is presented. Probenecid is the classical competitive inhibitor of organic acid transport in the kidney and other organs. There are 2 primary clinical
Wei Kong et al.
American journal of physiology. Heart and circulatory physiology, 307(1), H73-H79 (2014-05-06)
Previous experiments in cultures of neonatal rat myocytes demonstrated that the shape of Cai(2+) transients measured using high-affinity Ca(2+)-sensitive dyes may be misrepresented. The purpose of this study was to examine the role of dye affinity in Cai(2+) measurements in
R W Cowdry et al.
Biological psychiatry, 18(11), 1287-1299 (1983-11-01)
Probenecid is used to block the transport of acid monoamine metabolites from cerebrospinal fluid (CSF), on the assumption that the resultant rise in CSF concentrations of the metabolites will reflect presynaptic "turnover" of the parent monoamine. However, CSF levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service