Skip to Content
MilliporeSigma

1485001

USP

Oxybenzone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Hydroxy-4-methoxybenzophenone, Oxybenzone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOC6H3(OCH3)COC6H5
CAS Number:
Molecular Weight:
228.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1913145
eCl@ss:
39023842
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

oxybenzone

manufacturer/tradename

USP

bp

150-160 °C/5 mmHg (lit.)

mp

62-64 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc(c(O)c1)C(=O)c2ccccc2

InChI

1S/C14H12O3/c1-17-11-7-8-12(13(15)9-11)14(16)10-5-3-2-4-6-10/h2-9,15H,1H3

InChI key

DXGLGDHPHMLXJC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxybenzone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dioxybenzone and Oxybenzone Cream
  • Dong Quai Root
  • Dong Quai Root Powder
  • Oxybenzone
  • Sichuan Lovage Rhizome
  • Sichuan Lovage Rhizome Powder
  • St. John′s Wort Flowering Top

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Oxybenzone


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library