Skip to Content
MilliporeSigma

1477900

USP

Octinoxate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Ethylhexyl 4-methoxycinnamate, 4-Methoxycinnamic acid 2-ethylhexyl ester, OMC, Octyl methoxycinnamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H26O3
CAS Number:
Molecular Weight:
290.40
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5946632
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

octinoxate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(CC)COC(=O)\C=C\c1ccc(OC)cc1

InChI

1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3/b13-10+

InChI key

YBGZDTIWKVFICR-JLHYYAGUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Octinoxate USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare standard and system suitability solution during the assay of Octinoxate by using gas chromatography in conjunction with flame ionization detector.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Octinoxate


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

flash_point_f

379.4 °F

flash_point_c

193 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library