Skip to Content
MilliporeSigma

1477604

USP

Octreotide acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Octreotide acetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C49H66N10O10S2 · xC2H4O2
CAS Number:
Molecular Weight:
1019.24 (free base basis)
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

octreotide

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

S1SCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1)C(=O)N[C@@H]([C@H](O)C)CO)C(O)C)CCCCN)Cc4c5c([nH]c4)cccc5)Cc3ccccc3)NC(=O)C(N)Cc2ccccc2.OC(=O)C.OC(=O)C

InChI

1S/C49H66N10O10S2.2C2H4O2/c1-28(61)39(25-60)56-48(68)41-27-71-70-26-40(57-43(63)34(51)21-30-13-5-3-6-14-30)47(67)54-37(22-31-15-7-4-8-16-31)45(65)55-38(23-32-24-52-35-18-10-9-17-33(32)35)46(66)53-36(19-11-12-20-50)44(64)59-42(29(2)62)49(69)58-41;2*1-2(3)4/h3-10,13-18,24,28-29,34,36-42,52,60-62H,11-12,19-23,25-27,50-51H2,1-2H3,(H,53,66)(H,54,67)(H,55,65)(H,56,68)(H,57,63)(H,58,69)(H,59,64);2*1H3,(H,3,4)/t28-,29?,34?,36?,37?,38?,39-,40?,41?,42?;;/m1../s1

InChI key

QWFYIFWTVZFPRY-AARKYNAGSA-N

General description

Octreotide acetate is a synthetic cyclic octapeptide that mimics natural somatostatin, inhibiting the release of growth hormone, insulin, and glucagon. It is used therapeutically in acromegaly and neuroendocrine tumors. The USP reference standard enables identity, potency, and impurity testing in octreotide drug products.

The USP biologics peptides category encompasses a diverse range of therapeutic peptides that are essential in managing various medical conditions. These peptides, typically consisting of amino acid sequences of 40 residues or less, are critical for the development of high-quality medicines. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of peptide therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

This USP reference standard is intended for use in specified quality tests and assays.
Also used to prepare standard and system suitability solutions in the assay of octreotide acetate as described in the United States Pharmacopeia (USP) monograph.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.


Still not finding the right product?

Explore all of our products under Octreotide acetate


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library