Skip to Content
MilliporeSigma

Skip To

1475008

USP

Novobiocin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Streptonivicin, Benzamide, N-[7-[[3-O-(aminocarbonyl)-6-deoxy-5-C-methyl-4-O-methyl-β-l-lyxohexopyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-
(3-methyl-2-butenyl), monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H36N2O11 · H2O
CAS Number:
Molecular Weight:
630.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

novobiocin

form

solid

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Na+].N([C-]2C(=O)Oc3c(ccc(c3C)OC4OC(C(C(C4O)OC(=O)N)OC)(C)C)C2=O)C(=O)c1cc(c(cc1)O)CC=C(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Novobiocin is a coumarin antibiotic that inhibits bacterial DNA gyrase, thereby impeding DNA replication. It is used for selective microbial inhibition in culture media and research applications. The USP standard ensures the accuracy of chromatographic and bioassay-based quality evaluations.

The USP biologics antibiotics category includes a wide range of antimicrobial agents that are essential in treating bacterial infections. These antibiotics are derived from various natural sources or synthesized to combat specific pathogens effectively. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of antibiotic therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
136758113676061448978
grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

form

solid

form

-

form

-

form

-

format

neat

format

-

format

neat

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

novobiocin

API family

linezolid

API family

linezolid

API family

mycophenolate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters