Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1445620

USP

Moexipril Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-tert-Butyl 2-{(S)-2-[(S)-1-ethoxy-1-oxo-4-phenylbutan-2-ylamino]propanoyl}-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylate maleate, (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid 1,1-dimethylethyl ester (Z)-2-butenedioate (1:1), Moexipril tert-butyl ester maleic acid salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H42N2O7 · C4H4O4
CAS Number:
Molecular Weight:
670.75
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

moexipril

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N2([C@@H](Cc3c(cc(c(c3)OC)OC)C2)C(=O)OC(C)(C)C)C(=O)[C@@H](N[C@@H](CCc1ccccc1)C(=O)OCC)C.OC(=O)\C=C/C(=O)O

InChI

1S/C31H42N2O7.C4H4O4/c1-8-39-29(35)24(15-14-21-12-10-9-11-13-21)32-20(2)28(34)33-19-23-18-27(38-7)26(37-6)17-22(23)16-25(33)30(36)40-31(3,4)5;5-3(6)1-2-4(7)8/h9-13,17-18,20,24-25,32H,8,14-16,19H2,1-7H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t20-,24-,25-;/m0./s1

InChI key

SMLOIRKJVBQPQD-SAYMYIPQSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Moexipril Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Moexipril Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service