Skip to Content
MilliporeSigma

1445404

USP

Modafinil

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[(Diphenylmethyl)sulfinyl]­acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

modafinil

manufacturer/tradename

USP

drug control

USDEA Schedule IV; Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical (small molecule)

SMILES string

NC(=O)CS(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)

InChI key

YFGHCGITMMYXAQ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Modafinil is a central nervous system vigilance promoting agent, which posseses neuroprotective properties.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.


Still not finding the right product?

Explore all of our products under Modafinil


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library