Skip to Content
MilliporeSigma

1444300

USP

Misoprostol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H38O5
CAS Number:
Molecular Weight:
382.53
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

misoprostol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCC(C)(O)C\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)OC

InChI

1S/C22H38O5/c1-4-5-14-22(2,26)15-10-12-18-17(19(23)16-20(18)24)11-8-6-7-9-13-21(25)27-3/h10,12,17-18,20,24,26H,4-9,11,13-16H2,1-3H3/b12-10+/t17-,18-,20-,22?/m1/s1

InChI key

OJLOPKGSLYJEMD-URPKTTJQSA-N

Gene Information

human ... PTGER3(5733)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Ph

Application

Misoprostol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diclofenac Sodium and Misoprostol Delayed-Release Tablets
  • Misoprostol
  • Misoprostol Dispersion

Biochem/physiol Actions

PGE1 analog prodrug which is rapidly de-esterified to active "misoprostolic acid". Cited for extremely wide-ranging therapeutic effects, including prevention of NSAID-induced gastric ulceration, regulation of immunologic cascades, inhibition of platelet-activating factor (PAF), treatment of ethanol- and acetaminophen-induced hepatotoxicity and hepatitis, and stimulation of cartilage repair after injury.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Misoprostol


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library