Skip to Content
MilliporeSigma

1414025

USP

Methotrexate Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Aminopterin, (S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, 4-Amino-PGA, 4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H20N8O5
CAS Number:
Molecular Weight:
440.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Beilstein/REAXYS Number:
69045
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

methotrexate

manufacturer/tradename

USP

ε (extinction coefficient)

24,500 at 282 nm in 0.1 M NaOH at 1 M, 25,700 at 261 nm in 0.1 M NaOH at 1 M, 8,100 at 373 nm in 0.1 M NaOH at 1 M

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1nc(N)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)cnc2n1

InChI

1S/C19H20N8O5/c20-15-14-16(27-19(21)26-15)23-8-11(24-14)7-22-10-3-1-9(2-4-10)17(30)25-12(18(31)32)5-6-13(28)29/h1-4,8,12,22H,5-7H2,(H,25,30)(H,28,29)(H,31,32)(H4,20,21,23,26,27)/t12-/m0/s1

InChI key

TVZGACDUOSZQKY-LBPRGKRZSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methotrexate Related Compound B USP reference standard for use in specified quality tests and assays.

Additionally used to prepare standard, standard stock, system suitability, and system suitability stock solutions for impurity analysis in the given below monographs according to United States Pharmacopeia (USP):
  • Methotrexate
  • Methotrexate Injection
  • Methotrexate for Injection

Biochem/physiol Actions

Folic acid antagonist that binds to dihydrofolate reductase and inhibits its activity.
Folic acid antagonist. Aminopterin is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folylpolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Aminopterin-polyglutamate is degraded intracellularly by γ-glutamyl hydrolase.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

More potent, but more toxic, than methotrexate.
Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Methotrexate Related Compound B


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library