Skip to Content
MilliporeSigma

1379081

USP

Calcium D,L-5-methyltetrahydrofolate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-[4-[[(2-Amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H23CaN7O6
CAS Number:
Molecular Weight:
497.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

calcium d,l-5-methyltetrahydrofolate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Ca+2].N1(C(CNc3nc(nc(c31)[O-])N)CNc2ccc(cc2)C(=O)N[C@@H](CCC(=O)[O-])C(=O)O)C

InChI

1S/C20H25N7O6.Ca/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29;/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31);/q;+2/p-2/t12?,13-;/m0./s1

InChI key

VWBBRFHSPXRJQD-ZEDZUCNESA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This USP reference standard is intended for use in specified quality tests and assays.
Also used to prepare standard and system suitability solutions in the strength and impurity analysis by HPLC according to the following United States Pharmacopeia (USP) monographs:
  • Calcium l-5-Methyltetrahydrofolate Tablets
  • Calcium l-5-Methyltetrahydrofolate Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Calcium l-5-Methyltetrahydrofolate Capsules
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 4841-4841 (2016)
Calcium l-5-Methyltetrahydrofolate Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 4842-4842 (2016)



Global Trade Item Number

SKUGTIN
1379081-300MG04061833845547