Skip to Content
MilliporeSigma

Skip To

1359958

USP

Levocetirizine amide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(R)-2-(2-{4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl}ethoxy)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H26ClN3O2
CAS Number:
Molecular Weight:
387.90
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

levocetirizine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1359969BP837BP067
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

BP

application(s)

pharmaceutical (small molecule)
pharmaceutical

application(s)

pharmaceutical (small molecule)
pharmaceutical

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical
pharmaceutical small molecule

API family

levocetirizine

API family

levocetirizine

API family

cetirizine

API family

chlorcyclizine


Still not finding the right product?

Explore all of our products under Levocetirizine amide


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters