Skip to Content
MilliporeSigma
All Photos(1)

Documents

1351005

USP

Isoproterenol hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Isoprenaline hydrochloride, (±)-Isoproterenol hydrochloride, 1-(3′,4′-Dihydroxyphenyl)-2-isopropylaminoethanol hydrochloride, N-Isopropyl-DL-noradrenaline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72
Beilstein:
3917363
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isoproterenol

manufacturer/tradename

USP

mp

165-175 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)NCC(O)c1ccc(O)c(O)c1

InChI

1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H

InChI key

IROWCYIEJAOFOW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoproterenol hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Isoproterenol Hydrochloride Injection

Biochem/physiol Actions

β-adrenoceptor agonist; increases cytosolic cAMP.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoproterenol Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(4), 2431-2431 (2020)
Isoproterenol Hydrochloride Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(2), 2433-2433 (2021)
Hua Qing et al.
Cell, 182(2), 372-387 (2020-07-02)
Acute psychological stress has long been known to decrease host fitness to inflammation in a wide variety of diseases, but how this occurs is incompletely understood. Using mouse models, we show that interleukin-6 (IL-6) is the dominant cytokine inducible upon
Miaoying Shi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 67, 104891-104891 (2020-05-25)
Cardiotoxicity is an important toxicological endpoint for chemical and drug safety assessment. The present study aims to evaluate two stemcell-based in vitro models for cardiotoxicity screening of chemicals. Eleven model compounds were used to evaluate responses of mouse embryonic stem
Jason Z Zhang et al.
Cell, 182(6), 1531-1544 (2020-08-28)
The fidelity of intracellular signaling hinges on the organization of dynamic activity architectures. Spatial compartmentation was first proposed over 30 years ago to explain how diverse G protein-coupled receptors achieve specificity despite converging on a ubiquitous messenger, cyclic adenosine monophosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service