Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1287722

USP

Galactosamine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-(+)-Galactosamine hydrochloride, 2-Amino-2-deoxy-D-galactopyranose hydrochloride, D-Chondrosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
3697825
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

galactosamine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.N[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)CO

InChI

1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4+,5+,6-;/m0./s1

InChI key

CBOJBBMQJBVCMW-NQZVPSPJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Galactosamine hydrochloride USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Galactosamine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Heparin Sodium

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heparin Sodium
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(6), 2195-2195 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service