Skip to Content
MilliporeSigma

1286413

USP

Frovatriptan succinate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Frovatriptan succinate monohydrate, Frovatriptan succinate, R-(+) 3-Methylamino-6-carboxamido-1,2,3,4-tetrahydrocarbazole monosuccinate monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H17N3O · C4H6O4 · H2O
CAS Number:
Molecular Weight:
379.41
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

frovatriptan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(CCC(O)=O)=O.NC(C(C=C1)=CC2=C1NC3=C2C[C@H](NC)CC3)=O.O

InChI

1S/C14H17N3O.C4H6O4.H2O/c1-16-9-3-5-13-11(7-9)10-6-8(14(15)18)2-4-12(10)17-13;5-3(6)1-2-4(7)8;/h2,4,6,9,16-17H,3,5,7H2,1H3,(H2,15,18);1-2H2,(H,5,6)(H,7,8);1H2/t9-;;/m1../s1

InChI key

CUETXFMONOSVJA-KLQYNRQASA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Frovatriptan succinate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Frovatriptan is a serotonin 5HT-1B/1D-receptor agonist used to treat migraine. Frovatriptan has high affinity for the 5-HT(1B) and 5-HT(1D) receptors while affinity for 5-HT receptors other than 5-HT(1B/1D) is substantially lower with a pEC50 value of 8.2 for the 5-HT1B receptor, compared to a pEC50 value of 6.2 for the 5-HT7 receptor.
Frovatriptan is a serotonin 5HT-1B/1D-receptor agonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1286413-200MG04061833830802