Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1286060

USP

Formononetin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

7-Hydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 7-Hydroxy-3-(4-methoxyphenyl)chromone, 7-Hydroxy-4′-methoxyisoflavone, Biochanin B, Daidzein 4′-methyl ether, Formonetin, Formononetol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O4
CAS Number:
Molecular Weight:
268.26
Beilstein:
237979
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

formononetin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=COc3cc(O)ccc3C2=O

InChI

1S/C16H12O4/c1-19-12-5-2-10(3-6-12)14-9-20-15-8-11(17)4-7-13(15)16(14)18/h2-9,17H,1H3

InChI key

HKQYGTCOTHHOMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Isoflavone found in red clover. Effective against Giardia lamblia infection, at least partially by inducing detachment of trophozoites from intestinal mucosa.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service