Skip to Content
MilliporeSigma

Skip To

1270719

USP

Flavoxate Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

flavoxate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1c2c(cccc2C(=O)O)C(=O)C(=C1c3ccccc3)C

InChI

1S/C17H12O4/c1-10-14(18)12-8-5-9-13(17(19)20)16(12)21-15(10)11-6-3-2-4-7-11/h2-9H,1H3,(H,19,20)

InChI key

KMMBBZOSQNLLMN-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flavoxate Related Compound A USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as
  • Flavoxate Hydrochloride
  • Flavoxate Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR3317PHR35921270731
description

United States Pharmacopeia (USP) Reference Standard

description

certified reference material, pharmaceutical secondary standard

description

certified reference material, pharmaceutical secondary standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

agency

USP

agency

traceable to USP 1270719

agency

traceable to USP 1270731

agency

USP


Still not finding the right product?

Explore all of our products under Flavoxate Related Compound A


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters